missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Toluene, anhydrous, 99.8%, over molecular sieves, packaged under Argon in resealable ChemSeal™ bottles

Brand:  Alfa Aesar™ 047136.AE

Code : NEW

Additional Details : CAS Number : 108-88-3 Weight : 0.00010kg Transport : UN number : 1294 Chem class : 3 Pack group : II

Quantity 100mL

 View more versions of this product

Product Code. 15415075

  • 31.10 € / 100mL


Estimated Shipment date
from Supplier 23-08-2018
Add to basket

Description

Description

  • Packaged under Argon
Specifications

Chemical Identifiers

108-88-3
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
CHEBI:17578
C7H8
MFCD00008512
1140

Specifications

Toluene
108-88-3
C7H8
MFCD00008512
635760
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
1140
92.14
0.867g/mL
ChemSeal™ bottles
111°C
-93°C
Anhydrous, over molecular sieves
99.8%
C6H5CH3
UN1294
14,9529
CC1=CC=CC=C1
92.141
CHEBI:17578
Liquid
Petroleum-like
1.4967
Colorless
100mL
Videos
SDS