Learn More
Invitrogen™ TOP10 Electrocomp™ Kit
Beschreibung
TOP10 Electrocomp E. coli cells are provided at a transformation efficiency of 1 x 1010 cfu/μg supercoiled DNA and are ideal for high-efficiency cloning and plasmid propagation. This strain is used in wide range of applications and allows stable replication of high-copy number plasmids and maintenance of large plasmids.
The TOP10 genotype is similar to the DH10B strain and features:
• hsdR for efficient transformation of unmethylated DNA from PCR amplifications
• mcrA for efficient transformation of methylated DNA from genomic preparations
• lacZΔM15 for blue/white color screening of recombinant clones
• endA1 for cleaner preparations of DNA and better results in downstream applications due to the elimination of non-specific digestion by Endonuclease I
• recA1 for reduced occurrence of non-specific recombination in cloned DNA
Notes
Please see the product manual for reaction scale recommendations. A high-voltage electroporation apparatus is required.
Genotype
F- mcrA Δ(mrr-hsdRMS-mcrBC) φ80lacZΔM15 ΔlacX74 recA1 araD139 Δ(ara-leu)7697 galU galK rpsL endA1 nupG
Find the strain and format that fits your needs
• TOP10 is available in chemically competent and electrocompetent cell formats.
• Choose MultiShot competent cells for high-throughput cloning applications.
Spezifikation
Spezifikation
| Product Type | Electrocompetent Cells |
| Contains F' Episome | No |
| Improves Plasmid Quality | Yes (endA1) |
| Cloning Methylated DNA | Yes (mcrA) |
| Transformation Efficiency Level | High Efficiency (>1 x 1010 cfu/μg) |
| Content And Storage | • 5 x 100μL TOP10 Electrocomp E. coli • 50 μL pUC19 vector (10 pg/μL) Store at –80°C. • S.O.C. Medium (15 mL) Store at room temperature. |
| Antibiotic Resistance Bacterial | Yes (Streptomycin) |
| Cloning Unstable DNA | Not suitable for cloning unstable DNA |
| Blue/White Screening | Yes (lacΔM15) |
| High-throughput Compatibility | Low |
| Mehr anzeigen |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.