Learn More
Invitrogen™ Total Nitric Oxide Assay Kit
Measure total nitric oxide (NO) as the single value (sum) of nitrate (NO3-) and nitrite (NO2-) levels in serum, plasma, urine, saliva and other biological fluids.
Brand: Invitrogen™ EMSNOTOT
Code : A0
Additional Details : Weight : 0.10000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
Assay total nitric oxide (NO) as the single value (sum) of nitrate (NO3-) and nitrite (NO2-) levels in urine, saliva, plasma, serum and other biological fluids with our Thermo Scientific Total Nitric Oxide Assay Kit.
Additional Information and Online Ordering »
Target: Total nitric oxide (NO) contributed by the sum of nitrite (NO2-) and nitrate (NO3-)
Format: Colorimetric stain (azo dye) for clear 96-well plates
Measurement: Absorbance at 540nm (±20nm)
Assay Range: 3.1 to 100μM
Sensitivity: 1.35μM
Precision: Dependent upon several factors (generally, intra- and inter-assay CV ≤ 5%)
Sample Types: Urine, saliva, plasma, serum and other biological fluids
Total Assay Time: 45 minutes
Sample Size: 50μL
Includes:Complete kit contains Griess reagents, nitrite and nitrate standards, nitrate reductase, NADH, and diluents.
For Research Use Only. Not for use in diagnostic procedures.