Learn More
Toxicity Characteristic Leachate Procedure Standard 1, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep TCLP-100N
Code : NEW
Additional Details : CAS Number : 14797-71-8 Weight : 0.12500kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ba | 500μg/mL |
Ag | 25μg/mL |
As | 25μg/mL |
Cr | 25μg/mL |
Pb | 25μg/mL |
Cd | 5μg/mL |
Se | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
TCLP Standard 1 without Mercury | |
Triple-leached LDPE bottle | |
7440-39-3,7440-22-4,7440-38-2,7697-37-2,7732-18-5 | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Ba( 500mg/L),Ag(25mg/L),As(25mg/L) |
Without Mercury | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Assurance | |
2% HNO3 |
Product intended for laboratory use only