missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPM1 Recombinant Rabbit Monoclonal Antibody (K01_3K52)

Product Code. 30830352
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830352 50 μL
30827049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830352 Supplier Invitrogen™ Supplier No. MA565817

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TPM1 Recombinant Monoclonal Antibody for Western Blot

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPM1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K52
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene TPM1
Gene Accession No. P04692, P09493, P58771
Gene Alias AA986836; AI854628; alpha tropomyosin; alpha tropomyosin of brain; alpha tropomyosin of fibroblasts F2; alpha-fTM; Alpha-tm; alpha-Tm-1; alpha-Tm-2 isoform; alpha-tropomyosin; alpha-tropomyosin (partial); alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; C15orf13; cardiac alpha tropomyosin; cardiomyopathy, hypertrophic 3; CMD1Y; CMH3; epididymis secretory protein Li 265; HEL-S-265; hepatoma alpha tropomyosin; HTM-alpha; LVNC9; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; Tm1; TM2; Tm3; Tma2; Tmpa; Tmsa; Tpm1; Tpm-1; tpm1 protein; TPM1-1; TPM1-alpha; TPMA; tropomyosin; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1; tropomyosin 1 (alpha); tropomyosin 1 alpha chain; tropomyosin 1, alpha; tropomyosin 1-1; tropomyosin 3 alpha; tropomyosin alpha-1 chain; Tropomyosin-1; unnamed protein product; zgc:171719; zgc:55951; zgc:77009
Gene Symbols TPM1
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22003, 24851, 7168
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.