Learn More
Epredia™ TPX Single Sample Chamber, Caps and Filter Cards
For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
Brand:  Epredia™ A78710018
Code : 50
Additional Details : Weight : 0.22590kg
 View more versions of this product
Product Code. 12096669
  • 964.00 € / Pack of 12
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
TPX Single Sample Chamber
  • Reusable
  • Autoclavable
Filter Cards
  • Highly absorbent disposable filter cards for use with TPX chamber
  • Not for use with Cytofunnel™ disposable sample chambers
Specifications
Specifications
TPX Sample Chamber