Learn More
Fisherbrand™ Traceable™ Cradle and USB Cable for Logger-Trac™ Temp Mini Datalogger
Use this cradle and USB cord with the 6548 Mini-Logger-Trac Datalogger.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.05000kg
Product Code. 15547603
  • 75.35 € / Each
In Stock
to see stock.
Add to basket
Description
Description
  • This Cradle (6549) is a required accessory for the Fisherbrand™ Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer (6548)
  • Use for downloading temperature reading from Logger-Trac Datalogger
  • Cradle is constructed of plastic
  • Includes USB cable for downloading data onto datalogger
Specifications
Specifications
Cradle and USB Cable
Logger-Trac™ Temp Mini Datalogger