missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, ACROS Organics™

Brand:  Acros Organics 155000025

Code : NEW

Additional Details : CAS Number : 125572-95-4 Weight : 2.50000kg


Quantity 2.5kg

 View more versions of this product

Product Code. 15264644

  • 2462.30 € / 2.50kg

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

125572-95-4
364.35
VASZYFIKPKYGNC-DHTOPLTISA-N
2723844
C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O
C14H22N2O8·H2O
MFCD00149243
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate

Specifications

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate
0.3% max.
White
2.5kg
97.5% min. (Complexometry)
C6H10[N(CH2CO2H)2]2·H2O
13,III,1
Solubility in water: insoluble. Other solubilities: insoluble in most common organic solvents, soluble in alkali solutions
C1CCC(C(C1)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O
364.35
364.35
98%
Authentic
4.0 to 6.9% (K.F.)
>210.0°C
125572-95-4
C14H22N2O8·H2O
MFCD00149243
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd
VASZYFIKPKYGNC-DHTOPLTISA-N
2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate
2723844
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Documents