missing translation for 'onlineSavingsMsg'
Learn More

trans-2-Pentenoic acid, 90+%, remainder other isomers, Thermo Scientific Chemicals

Catalog Number p-7020656
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
13991-37-2
C5H8O2
100.12
MFCD00002704
YIYBQIKDCADOSF-ONEGZZNKSA-N
trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
638122
CHEBI:38366
(E)-pent-2-enoic acid
CC\C=C\C(O)=O
This item is not returnable. View return policy
13991-37-2
C5H8O2
100.12
MFCD00002704
YIYBQIKDCADOSF-ONEGZZNKSA-N
trans-2-pentenoic acid, 2-pentenoic acid, e-pent-2-enoic acid, pent-2-enoic acid, 2e-pent-2-enoic acid, 2-pentenoic acid, 2e, beta-ethyl acrylic acid, c2h5ch=chcooh, pentenoic acid, unii-1rg66883cf
638122
CHEBI:38366
(E)-pent-2-enoic acid
CC\C=C\C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 8°C to 10°C
Density 0.992
Boiling Point 200°C to 203°C
Flash Point 102°C (215°F)
Odor Pungent
Refractive Index 1.4515
Quantity 10 g
UN Number UN3265
Beilstein 1720312
Formula Weight 100.12
Percent Purity ≥90%
Assay remainder other isomers
Chemical Name or Material trans-2-Pentenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'