missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Hexenoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7021195
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
1577-18-0
C6H10O2
114.14
MFCD00002786
XXHDAWYDNSXJQM-ONEGZZNKSA-N
trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
5282708
CHEBI:49285
(E)-hex-3-enoic acid
CC\C=C\CC(O)=O
This item is not returnable. View return policy
1577-18-0
C6H10O2
114.14
MFCD00002786
XXHDAWYDNSXJQM-ONEGZZNKSA-N
trans-3-hexenoic acid, hydrosorbic acid, 3-hexenoic acid, e-hex-3-enoic acid, 3-hexenoic acid, 3e, e-3-hexenoic acid, 3-hexenoic acid, e, trans-hex-3-enoic acid, unii-9b3n069edg, fema no. 3170
5282708
CHEBI:49285
(E)-hex-3-enoic acid
CC\C=C\CC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 11°C to 12°C
Density 0.961
Boiling Point 206°C
Flash Point 99°C (210°F)
Odor Rancid
Refractive Index 1.4398
Quantity 10 g
UN Number UN3265
Beilstein 1700862
Formula Weight 114.14
Percent Purity 97%
Chemical Name or Material trans-3-Hexenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'