missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline, 97%

Brand:  Alfa Aesar™ H61194.03

Code : NEW

Additional Details : CAS Number : 96034-57-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15476336

  • 39.60 € / 1g


Estimated Shipment date
from Supplier 23-05-2019
Add to basket

Specifications

Chemical Identifiers

96034-57-0
310.262
trans-4-hydroxy-1-4-nitrobenzyloxycarbonyl-l-proline, 2s,4r-4-hydroxy-1,2-pyrrolidinedicarboxylic acid 1-4-nitrobenzyl ester, 2s,4r-4-hydroxy-1,2-pyrrolidinedicarboxylic acid 1-4-nitrobenzyl ester, 2s,4r-1-4-nitrobenzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4r-4-hydroxy-1-4-nitrophenyl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-p-nitrobenzyloxycarbonyl-l-hydroxyproline, trans-1-p-nitrobenzyloxycarbonyl-4-hydroxy-l-proline, 2s,4r-1-p-nitrobenzyloxycarbonyl-4-hydroxyproline, 2s,4r-4-hydroxy-1-4-nitrobenzyloxycarbonyl proline, 1-p-nitrobenzyloxycarbonyl-trans-4-hydroxy-l-proline
(2S,4R)-4-hydroxy-1-[(4-nitrophenyl)methoxycarbonyl]pyrrolidine-2-carboxylic acid
C13H14N2O7
JMJMJDNHVXYAOC-MNOVXSKESA-N
10566895
C1C(CN(C1C(=O)O)C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])O

Specifications

trans-4-Hydroxy-1-(4-nitrobenzyloxycarbonyl)-L-proline
C13H14N2O7
JMJMJDNHVXYAOC-MNOVXSKESA-N
(2S,4R)-4-hydroxy-1-[(4-nitrophenyl)methoxycarbonyl]pyrrolidine-2-carboxylic acid
10566895
Solid
1g
96034-57-0
trans-4-hydroxy-1-4-nitrobenzyloxycarbonyl-l-proline, 2s,4r-4-hydroxy-1,2-pyrrolidinedicarboxylic acid 1-4-nitrobenzyl ester, 2s,4r-4-hydroxy-1,2-pyrrolidinedicarboxylic acid 1-4-nitrobenzyl ester, 2s,4r-1-4-nitrobenzyloxy carbonyl-4-hydroxypyrrolidine-2-carboxylic acid, 2s,4r-4-hydroxy-1-4-nitrophenyl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-p-nitrobenzyloxycarbonyl-l-hydroxyproline, trans-1-p-nitrobenzyloxycarbonyl-4-hydroxy-l-proline, 2s,4r-1-p-nitrobenzyloxycarbonyl-4-hydroxyproline, 2s,4r-4-hydroxy-1-4-nitrobenzyloxycarbonyl proline, 1-p-nitrobenzyloxycarbonyl-trans-4-hydroxy-l-proline
C1C(CN(C1C(=O)O)C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])O
310.262
310.26
97%
Videos
SDS