missing translation for 'onlineSavingsMsg'
Learn More

trans-Stilbene, 96%, Thermo Scientific Chemicals

Catalog Number p-193606
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
103-30-0
C14H12
180.25
MFCD00064300
PJANXHGTPQOBST-VAWYXSNFSA-N
trans-stilbene, e-stilbene, stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, bibenzylidene, bibenzylidine, bibenzal, trans-1,2-diphenylethene, alpha,beta-diphenylethylene
638088
CHEBI:36007
(E)-stilbene
C1=CC=C(C=C1)C=CC2=CC=CC=C2

trans-Stilbene, 96%, C14H12, CAS Number-103-30-0, bibenzylidine, trans-stilbene, 1,2-diphenylethylene, trans-1,2-diphenylethylene, alpha,beta-diphenylethylene, stilbene, trans-1,2-diphenylethene, bibenzal, e-stilbene, bibenzylidene, 500g, 95% min. (HPLC), 05,II,537

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 122°C to 126°C
Color White to Beige
Boiling Point 305°C to 307°C (744.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CH=CHC6H5
Quantity 100 g
Beilstein 05,II,537
Merck Index 15,8946
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in 90 parts cold alcohol,freely soluble in benzene and ether
Formula Weight 180.25
Percent Purity 96%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material trans-Stilbene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • trans-Stilbene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'