missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ALK tyrosine kinase receptor / CD246 siRNA
Blocks transcription and translation of ALK tyrosine kinase receptor / CD246 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_004295 |
---|---|
Cell Lines Tested | SK-N-MC and NB-39-nu Neuroblastoma Cell Lines. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | ALK |
Gene ID (Entrez) | 238 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059161
|
enQuireBio™
QX1-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049161
|
enQuireBio™
QX1-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004295 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
238 | |
CD246, NBLST3, Anaplastic lymphoma kinase | |
Lyophilized |
SK-N-MC and NB-39-nu Neuroblastoma Cell Lines. | |
ALK | |
siRNA | |
Human |