missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Alpha-synuclein siRNA
Blocks transcription and translation of Alpha-synuclein from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000336 |
---|---|
Cell Lines Tested | SH-SY5Y. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | SNCA |
Gene ID (Entrez) | 6622 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059181
|
enQuireBio™
QX2-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049181
|
enQuireBio™
QX2-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000336 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
6622 | |
PD1, NACP, PARK1, PARK4, SNCA | |
Lyophilized |
SH-SY5Y. | |
SNCA | |
siRNA | |
Human |