missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Amyloid beta A4 protein siRNA
Blocks transcription and translation of Amyloid beta A4 protein from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000475 |
---|---|
Cell Lines Tested | HEK 293. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | APP |
Gene ID (Entrez) | 351 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079201
|
enQuireBio™
QX3-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069201
|
enQuireBio™
QX3-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000475 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
351 | |
CTFgamma, ABETA, AD1, PN2, ABPP, CVAP, AAA, APPI, PN-II, ab | |
Lyophilized |
HEK 293. | |
APP | |
siRNA | |
Human |