missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Androgen receptor siRNA
Blocks transcription and translation of Androgen receptor from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000035 |
---|---|
Cell Lines Tested | LNCaP and LNCaP-Rf Androgen Dependent and Independent Metastatic Cells.. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | AR |
Gene ID (Entrez) | 367 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099221
|
enQuireBio™
QX4-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089221
|
enQuireBio™
QX4-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000035 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
367 | |
SMAX1, AR8, KD, SBMA, HUMARA, DHTR, HYSP1, TFM, AIS, NR3C4 | |
Lyophilized |
LNCaP and LNCaP-Rf Androgen Dependent and Independent Metastatic Cells.. | |
AR | |
siRNA | |
Human |