missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ApoE siRNA
Blocks transcription and translation of ApoE from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000032 |
---|---|
Cell Lines Tested | Ovcar3 Human Ovarian Cancer Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | APOE |
Gene ID (Entrez) | 348 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019251
|
enQuireBio™
QX5-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009251
|
enQuireBio™
QX5-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000032 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
348 | |
AD2, APO-E, LPG, LDLCQ5, ApoE4, ApoE, Apolipoprotein E | |
Lyophilized |
Ovcar3 Human Ovarian Cancer Cells. | |
APOE | |
siRNA | |
Human |