missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-1 siRNA
Blocks transcription and translation of Caspase-1 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_001214 |
---|---|
Cell Lines Tested | Human Primary Keratinocytes. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CASP1 |
Gene ID (Entrez) | 834 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049221
|
enQuireBio™
QX38-1NMOL |
1 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059221
|
enQuireBio™
QX38-5NMOL |
5 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001214 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
834 | |
Interleukin 1, Beta, Convertase, ICE, Caspase 1 | |
Lyophilized |
Human Primary Keratinocytes. | |
CASP1 | |
siRNA | |
Human |