missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-2 siRNA
Blocks transcription and translation of Caspase-2 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_116765 |
---|---|
Cell Lines Tested | U-2 OS Human Osteosarcoma Epithelial Cells from Bone. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CASP2 |
Gene ID (Entrez) | 835 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079221
|
enQuireBio™
QX39-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069221
|
enQuireBio™
QX39-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_116765 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
835 | |
CASP-2, ICH1, PPP1R57, NEDD-2, NEDD2, CASP2 | |
Lyophilized |
U-2 OS Human Osteosarcoma Epithelial Cells from Bone. | |
CASP2 | |
siRNA | |
Human |