missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Caspase-8 siRNA
Blocks transcription and translation of Caspase-8 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_001219 |
---|---|
Cell Lines Tested | 293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CASP8 |
Gene ID (Entrez) | 841 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079231
|
enQuireBio™
QX43-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069231
|
enQuireBio™
QX43-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001219 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
841 | |
CAP4, MCH5, FLICE, MACH, Casp-8, ALPS2B, CASP8 | |
Lyophilized |
293 Human Embryonic Kidney Cells with NF-kappa B and Flag tagged DR4 constructs. | |
CASP8 | |
siRNA | |
Human |