missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Cellular tumor antigen p53 siRNA
Blocks transcription and translation of Cellular tumor antigen p53 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000537 |
---|---|
Cell Lines Tested | U-2 OS Huan Osteosarcoma Epithelial Cells from Bone. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TP53 |
Gene ID (Entrez) | 7157 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039211
|
enQuireBio™
QX32-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029211
|
enQuireBio™
QX32-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000537 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7157 | |
TRP53, BCC7, P53, LFS1, TP53 | |
Lyophilized |
U-2 OS Huan Osteosarcoma Epithelial Cells from Bone. | |
TP53 | |
siRNA | |
Human |