missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human CTLA4 siRNA
Blocks transcription and translation of CTLA4 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_005205 |
---|---|
Cell Lines Tested | CLL Chronic Lymphoid Leukemia Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | CTLA4 |
Gene ID (Entrez) | 1493 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059241
|
enQuireBio™
QX47-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049241
|
enQuireBio™
QX47-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005205 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
1493 | |
Cytotoxic T-Lymphocyte Associated Protein 4, CTLA-3, CD152, Celiac3, Celiac Disease 3, Cytotoxic T-Lymphocyte-Associated Antigen 4 | |
Lyophilized |
CLL Chronic Lymphoid Leukemia Cells. | |
CTLA4 | |
siRNA | |
Human |