missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human ErbB4 / Her4 siRNA
Blocks transcription and translation of ErbB4 / Her4 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_005226 |
---|---|
Cell Lines Tested | T-47d Human Carcinoma Cells from Breast. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | ERBB4 |
Gene ID (Entrez) | 2066 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099181
|
enQuireBio™
QX21-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089181
|
enQuireBio™
QX21-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005226 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2066 | |
p180erbB4, HER4, ALS19, Erb-b4, ErbB-4 | |
Lyophilized |
T-47d Human Carcinoma Cells from Breast. | |
ERBB4 | |
siRNA | |
Human |