missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor beta siRNA
Blocks transcription and translation of Estrogen receptor beta from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_034287 |
---|---|
Cell Lines Tested | MCF7 Human Adenocarcinoma Cells from Breast Cancer. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | ESR2 |
Gene ID (Entrez) | 13983 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079171
|
enQuireBio™
QX16-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069171
|
enQuireBio™
QX16-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_034287 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
13983 | |
ESR-BETA, NR3A2, ESTRB, Erb, ESRB, ER-BETA, ERBeta | |
Lyophilized |
MCF7 Human Adenocarcinoma Cells from Breast Cancer. | |
ESR2 | |
siRNA | |
Human |