missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Estrogen receptor siRNA
Blocks transcription and translation of Estrogen receptor from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000116 |
---|---|
Cell Lines Tested | MCF7 Human Adenocarcinoma Cells from Breast Cancer. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | ESR1 |
Gene ID (Entrez) | 2099 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039171
|
enQuireBio™
QX14-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029171
|
enQuireBio™
QX14-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000116 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
2099 | |
ESRA, ESR, NR3A1, ER, ESTRR, Era | |
Lyophilized |
MCF7 Human Adenocarcinoma Cells from Breast Cancer. | |
ESR1 | |
siRNA | |
Human |