missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human FoxP3 siRNA
Blocks transcription and translation of FoxP3 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_001107849 |
---|---|
Cell Lines Tested | HCT116-p53wt Human Carcinoma Cells from Colon and MCF7 Human Adenocarcinoma Cells from Breast Cancer. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | FoxP3 |
Gene ID (Entrez) | 50943 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079241
|
enQuireBio™
QX48-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069241
|
enQuireBio™
QX48-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001107849 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
50943 | |
Forkhead Box P3, Scurfin, IPEX | |
Lyophilized |
HCT116-p53wt Human Carcinoma Cells from Colon and MCF7 Human Adenocarcinoma Cells from Breast Cancer. | |
FoxP3 | |
siRNA | |
Human |