missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human HIF-1 alpha siRNA
Blocks transcription and translation of HIF-1 alpha from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_001521 |
---|---|
Cell Lines Tested | Human mda-mb-435 Breast Cancer Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | HIF1A |
Gene ID (Entrez) | 3091 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019171
|
enQuireBio™
QX12-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009171
|
enQuireBio™
QX12-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001521 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3091 | |
HIF-1-alpha, HIF1-ALPHA, HIF1, HIF-1alpha, PASD8, MOP1, HIF-1A, bHLHe78, Hypoxia-inducible factor 1-alpha | |
Lyophilized |
Human mda-mb-435 Breast Cancer Cells. | |
HIF1A | |
siRNA | |
Human |