missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Insulin Receptor siRNA
Blocks transcription and translation of Insulin Receptor from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000199 |
---|---|
Cell Lines Tested | HepG2 Human Hepatocellular Carcinoma Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Insr |
Gene ID (Entrez) | 3643 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059251
|
enQuireBio™
QX51-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049251
|
enQuireBio™
QX51-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000199 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3643 | |
InsR, IR, CD220 | |
Lyophilized |
HepG2 Human Hepatocellular Carcinoma Cells. | |
Insr | |
siRNA | |
Human |