missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Interleukin-10 / IL-10 siRNA
Blocks transcription and translation of Interleukin-10 / IL-10 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000563 |
---|---|
Cell Lines Tested | Dendritic Cells derived from adherent human PBMCs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | IL10 |
Gene ID (Entrez) | 3586 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019221
|
enQuireBio™
QX36-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009221
|
enQuireBio™
QX36-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000563 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3586 | |
IL10A, IL-10, TGIF, CSIF, GVHDS | |
Lyophilized |
Dendritic Cells derived from adherent human PBMCs. | |
IL10 | |
siRNA | |
Human |