missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human IRAK4 siRNA
Blocks transcription and translation of IRAK4 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_057207 |
---|---|
Cell Lines Tested | 293T Human Embryonic Kidney Cells with Large T Antigen. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | IRAK4 |
Gene ID (Entrez) | 51135 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059201
|
enQuireBio™
QX29-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049201
|
enQuireBio™
QX29-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057207 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
51135 | |
Interleukin-1 receptor-associated kinase 4, IRAK-4 | |
Lyophilized |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
IRAK4 | |
siRNA | |
Human |