missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human PD-L2 / CD273 siRNA
Blocks transcription and translation of PD-L2 / CD273 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_079515 |
---|---|
Cell Lines Tested | Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | PDCD1LG2 |
Gene ID (Entrez) | 80380 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019261
|
enQuireBio™
QX8-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009261
|
enQuireBio™
QX8-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_079515 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
80380 | |
PDL2, CD273, Programmed Cell Death 1 Ligand 2, PDCD1L2, B7-DC, B7DC, PD-1 L2, PD-1 Ligand 2, PDCD1 Ligand 2 | |
Lyophilized |
Hep3b Human Hepatocellular Carinoma Cells, RKO Human Colon Cancer Cells, and Primary Dendritic Cells (DCs) isolated from PBMCs. | |
PDCD1LG2 | |
siRNA | |
Human |