missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR3 siRNA
Blocks transcription and translation of TLR3 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_003256.1 |
---|---|
Cell Lines Tested | A549 Human Adenocarcinoma cells from Basal Alveolar Epithelium. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TLR3 |
Gene ID (Entrez) | 7098 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019211
|
enQuireBio™
QX31-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009211
|
enQuireBio™
QX31-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003256.1 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7098 | |
TLR-3, Toll-like Receptor 3, CD283 | |
Lyophilized |
A549 Human Adenocarcinoma cells from Basal Alveolar Epithelium. | |
TLR3 | |
siRNA | |
Human |