missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TLR7 / CD287 siRNA
Blocks transcription and translation of TLR7 / CD287 from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_057646 |
---|---|
Cell Lines Tested | 293T Human Embryonic Kidney Cells with Large T Antigen. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TLR7 |
Gene ID (Entrez) | 51284 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099191
|
enQuireBio™
QX26-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089191
|
enQuireBio™
QX26-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_057646 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
51284 | |
TLR7-like, TLR7, Toll-like receptor 7, TLR-7 | |
Lyophilized |
293T Human Embryonic Kidney Cells with Large T Antigen. | |
TLR7 | |
siRNA | |
Human |