missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human TNF alpha siRNA
Blocks transcription and translation of TNF alpha from Human.
250.00 € - 369.00 €
Specifications
Accession Number | NP_000585 |
---|---|
Cell Lines Tested | Adherent LPS Treated Human PBMCs and Adherent dsRNA Treated Human PBMCs. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | TNF |
Gene ID (Entrez) | 7124 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16019191
|
enQuireBio™
QX22-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009191
|
enQuireBio™
QX22-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000585 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
7124 | |
DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor | |
Lyophilized |
Adherent LPS Treated Human PBMCs and Adherent dsRNA Treated Human PBMCs. | |
TNF | |
siRNA | |
Human |