missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Mouse Apoptosis regulator BAX siRNA
Blocks transcription and translation of Apoptosis regulator BAX from Mouse.
250.00 € - 369.00 €
Specifications
Accession Number | NP_031553 |
---|---|
Cell Lines Tested | DC-1 Mouse Dendritic Cells. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Bax |
Gene ID (Entrez) | 12028 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039261
|
enQuireBio™
QX9-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029261
|
enQuireBio™
QX9-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031553 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
12028 | |
BCL2L4 | |
Lyophilized |
DC-1 Mouse Dendritic Cells. | |
Bax | |
siRNA | |
Mouse |