missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Mouse TNF alpha siRNA
Blocks transcription and translation of TNF alpha from Mouse.
250.00 € - 369.00 €
Specifications
Accession Number | NP_038721 |
---|---|
Cell Lines Tested | Murine Peritoneal Macrophage. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Tnf |
Gene ID (Entrez) | 21926 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039191
|
enQuireBio™
QX23-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029191
|
enQuireBio™
QX23-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_038721 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
21926 | |
DIF, TNFA, TNFSF2, TNLG1F, TNF-alpha, Tumor necrosis factor alpha, Tumor necrosis factor | |
Lyophilized |
Murine Peritoneal Macrophage. | |
Tnf | |
siRNA | |
Mouse |