missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Rat High Affinity Nerve Growth Factor Receptor (TrkA) siRNA
Blocks transcription and translation of High Affinity Nerve Growth Factor Receptor (TrkA) from Rat.
250.00 € - 369.00 €
Specifications
Accession Number | NP_067600 |
---|---|
Cell Lines Tested | Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. |
Content And Storage | Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. |
Gene Symbol | Ntrk1 |
Gene ID (Entrez) | 59109 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16079211
|
enQuireBio™
QX34-2NMOL |
2 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069211
|
enQuireBio™
QX34-10NMOL |
10 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_067600 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
59109 | |
TrkA, High Affinity NGFR, NGFR, Trk | |
Lyophilized |
Pc12 Rat Pheocromocytoma Cells from the Adrenal Medulla and Rat Primary Neurons. | |
Ntrk1 | |
siRNA | |
Rat |