missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Tray Support for Gel Stainer and Processor Plus
313.14 € - 455.94 €
Specifications
For Use With (Equipment) | Gel Stainer and Processor Plus, Blot Processor |
---|---|
Type | Tray Support |
Product Code | Brand | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||||
10026134
|
Cytiva
80-6344-29 |
4.22 oz. | 125 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11570904
|
Cytiva
80-6345-05 |
8.45 oz. | 250 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Specifications
Gel Stainer and Processor Plus, Blot Processor | |
Tray Support |