Learn More
Invitrogen™ Human TREM1, Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP75730
Code : LD
Additional Details : CAS Number : 69-74-9 Weight : 0.01000kg
Description
TREM1 (human) Recombinant Protein, RP-75730, from Invitrogen.
RP-75730 contains purified recombinant human TReM1 protein with an N-terminal hexahistidine tag. RP-75730 may be used as a positive control in many immunological assays.Specifications
Q9NP99 | |
Control, Western Blot | |
54210 | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Soluble Triggering Receptor Expressed on Myeloid Cells-1 | |
TREM1 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.05mg/mL | |
Laemmli with no preservative | |
Human TREM1 | |
E. Coli | |
RUO | |
Human TREM1 | |
Human | |
Hu | |
Recombinant | |
Recombinant |