missing translation for 'onlineSavingsMsg'
Learn More

Tri-Potassium Citrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

Catalog Number 10428610
Click to view available options
Quantity:
500 g
3 kg
This item is not returnable. View return policy
6100-05-6
C6H7K3O8
324.41
150442
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
This item is not returnable. View return policy
6100-05-6
C6H7K3O8
324.41
150442
PJAHUDTUZRZBKM-UHFFFAOYSA-K
potassium citrate monohydrate, tripotassium citrate monohydrate, unii-ee90oni6ff, ee90oni6ff, citric acid tripotassium salt, urocit-k, ccris 6543, tri-potassium citrate monohydrate, potassium citrate tribasic monohydrate, citric acid, tripotassium salt, monohydrate
2735208
CHEBI:64746
tripotassium;2-hydroxypropane-1,2,3-tricarboxylate;hydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.[K+].[K+].[K+]
Melting Point 180°C
Color White
pH 7.5 to 9.0
Physical Form Solid
Quantity 500 g
Formula Weight 324.42g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'