missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, 99%, Thermo Scientific Chemicals

Catalog Number p-4509960
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
102-76-1
C9H14O6
218.21
URAYPUMNDPQOKB-UHFFFAOYSA-N
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
5541
CHEBI:9661
2,3-diacetyloxypropyl acetate
CC(=O)OCC(COC(=O)C)OC(=O)C
102-76-1
C9H14O6
218.21
URAYPUMNDPQOKB-UHFFFAOYSA-N
triacetin, glyceryl triacetate, glycerol triacetate, glycerin triacetate, enzactin, triacetine, triacetylglycerol, fungacetin, glyped, triacetyl glycerine
5541
CHEBI:9661
2,3-diacetyloxypropyl acetate
CC(=O)OCC(COC(=O)C)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3.0°C
Color Colorless
Density 1.1550g/mL
Boiling Point 258.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3CO2CH2)2CH(O2CCH3)
Refractive Index 1.4290 to 1.4330
Quantity 500 mL
Beilstein 02,147
Merck Index 15,9751
Specific Gravity 1.155
Solubility Information Solubility in water: 64.0g/L (20°C). Other solubilities: soluble in most common organic solvents,slightly soluble in carbon disulfide
Viscosity 23 mPa.s (20°C)
Formula Weight 218.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triacetin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'