missing translation for 'onlineSavingsMsg'
Learn More

Tributyl borate, 98%, Thermo Scientific Chemicals

Catalog Number 11945161
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
688-74-4
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC
This item is not returnable. View return policy
688-74-4
LGQXXHMEBUOXRP-UHFFFAOYSA-N
n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
12712
tributyl borate
B(OCCCC)(OCCCC)OCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -70.0°C
Color Colorless
Density 0.8530g/mL
Boiling Point 230.0°C to 235.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H27BO3
Linear Formula (CH3CH2CH2CH2O)3B
Refractive Index 1.4080 to 1.4100
Quantity 2.5 L
Beilstein 01,II,398
Fieser 13,326
Specific Gravity 0.853
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with most common organic liquids
Molecular Weight (g/mol) 230.15
Formula Weight 230.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tributyl borate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'