missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine hydrochloride, +99%, Thermo Scientific Chemicals

Catalog Number p-203800
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
637-39-8
C6H16ClNO3
185.648
MFCD00012596
HHLJUSLZGFYWKW-UHFFFAOYSA-N
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
101814
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
C(CO)N(CCO)CCO.Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
637-39-8
C6H16ClNO3
185.648
MFCD00012596
HHLJUSLZGFYWKW-UHFFFAOYSA-N
triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
101814
2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
C(CO)N(CCO)CCO.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Loss on Drying 1.0% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (HOCH2CH2)3N·HCl
Quantity 5 g
Formula Weight 185.65
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material Triethanolamine hydrochloride, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'