missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog Number p-4509978
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
867-13-0
C8H17O5P
224.19
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC
867-13-0
C8H17O5P
224.19
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Acidity 2mg KOH/g max.
Melting Point -24.0°C
Density 1.1300g/mL
Boiling Point 260.0°C to 262.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
Refractive Index 1.4300 to 1.4320
Quantity 100 g
Beilstein 04,I,573
Fieser 01,1216; 06,612; 12,527; 13,255
Specific Gravity 1.13
Solubility Information Solubility in water: insoluble
Formula Weight 224.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Triethyl phosphonoacetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'