missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylaluminium, 1.3M solution in Heptane, Thermo Scientific Chemicals
Click to view available options
Quantity:
800 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
Specifications
Color | Colorless |
Linear Formula | (C2H5)3Al |
Concentration | 19.5 to 20.5 wt% (on Al basis) |
Density | 0.7300 g/mL |
Molecular Weight (g/mol) | 114.17 |
Formula Weight | 114.17 |
Packaging | Glass bottle |
Quantity | 800 mL |
Physical Form | Solution |
Chemical Name or Material | Triethylaluminium, 1.3 M Solution in Heptane |
Safety and Handling
Product Identifier
- Triethylaluminium, 1.M solution in heptane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Pyrophoric liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
- H304-May be fatal if swallowed and enters airways.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H250-Catches fire spontaneously if exposed to air.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'