Learn More
Triethylamine, for HPLC, Fisher Chemical™
C6H15N, CAS Number-121-44-8, 250mL, 90 deg.C, CHEBI:35026, Colorless, 101.19g/mol, ZMANZCXQSJIPKH-UHFFFAOYSA-N, N,N-diethylethanamine, -115 deg.C, 9051, 101.193, Amber glass bottle, 12.4, Liquid, 8471, CCN(CC)CC, 3.5, 69mbar at 20 deg.C, 0.36 mPaS at 20 deg.C
Brand: Fisher Chemical T/3203/07
Code : 25
Additional Details : CAS Number : 121-44-8 Weight : 0.18200kg Transport : UN number : 1296 Chem class : 3 Pack group : II
Description
C6H15N, CAS Number-121-44-8, 250mL, 90 deg.C, CHEBI:35026, Colorless, 101.19g/mol, ZMANZCXQSJIPKH-UHFFFAOYSA-N, N,N-diethylethanamine, -115 deg.C, 9051, 101.193, Amber glass bottle, 12.4, Liquid, 8471, CCN(CC)CC, 3.5, 69mbar at 20 deg.C, 0.36 mPaS at 20 deg.C
Specifications
Liquid | |
C6H15N | |
ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
N,N-diethylethanamine | |
8471 | |
101.19g/mol | |
69mbar at 20°C | |
3.5 | |
-115°C | |
Colorless |
121-44-8 | |
9051 | |
CCN(CC)CC | |
101.193 | |
CHEBI:35026 | |
Amber glass bottle | |
0.36 mPaS at 20°C | |
12.4 | |
90°C | |
250 mL |
Safety and Handling
- Acetic acid, sodium salt trihydrate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.