Learn More
Triethyl orthoformate, 98%, anhydrous, AcroSeal™, ACROS Organics™
Brand: Acros Organics 429278000
Code : 29
Additional Details : CAS Number : 122-51-0 Weight : 0.76200kg Transport : UN number : 2524 Chem class : 3 Pack group : III
Packaging | AcroSeal™ Glass Bottle |
---|---|
Quantity | 800mL |
Chemical Identifiers
122-51-0 | |
148.2 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
diethoxymethoxyethane |
C7H16O3 | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
31214 | |
CCOC(OCC)OCC |
Specifications
Triethyl orthoformate | |
35°C | |
1.3900 to 1.3920 | |
146.0°C | |
800mL | |
97.5% min. (GC) | |
CH(OC2H5)3 | |
triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
GKASDNZWUGIAMG-UHFFFAOYSA-N | |
diethoxymethoxyethane | |
31214 | |
98% |
0.8900g/mL | |
AcroSeal™ Glass Bottle | |
0.89 | |
-61.0°C | |
122-51-0 | |
C7H16O3 | |
15,698 | |
Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform, | |
CCOC(OCC)OCC | |
148.2 | |
148.2 |
Safety and Handling
- Triethyl orthoformate
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Sicherheitshinweis
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.