missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilane, 99%, Thermo Scientific Chemicals

Catalog Number p-4509581
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
617-86-7
C6H16Si
116.28
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC
This item is not returnable. View return policy
617-86-7
C6H16Si
116.28
QXTIBZLKQPJVII-UHFFFAOYSA-N
triethylsilane, hydride, silane, triethyl, triethylhydrosilane, triethylsilyl, silane e3h, c6h16si, triethylsilyl radical, silane, triethyl-,, triethylsilane tes
6327258
triethylsilicon
CC[Si](CC)CC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Boiling Point 107.0°C to 108.0°C
Color Colorless
Physical Form Liquid
Quantity 5 g
Assay Percent Range 99%
Linear Formula (C2H5)3SiH
Beilstein 04,625
Fieser 01,1218; 02,433; 03,304; 04,530; 05,694; 12,482; 13,340; 14,322; 15,338; 16,356; 17,367
Flash Point −3°C
Merck Index 15,9841
Solubility Information Solubility in water: decomposes
Formula Weight 116.28
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4100 to 1.4120
Specific Gravity 0.72
Density 0.7200g/mL
Chemical Name or Material Triethylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylsilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'