missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl trifluoromethanesulfonate, 99%, ACROS Organics™

Brand:  Acros Organics 312700500

Code : 29

Additional Details : CAS Number : 79271-56-0 Weight : 0.10000kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 11975831

  • 167.00 € / 50g

Estimated Shipment
27-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Silylation reagent
Specifications

Chemical Identifiers

79271-56-0
264.34
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
triethylsilyl trifluoromethanesulfonate
C7H15F3O3SSi
STMPXDBGVJZCEX-UHFFFAOYSA-N
2733308
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F

Specifications

Triethylsilyl trifluoromethanesulfonate
72°C
Glass bottle
1.16
Colorless to Brown
79271-56-0
C7H15F3O3SSi
triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
STMPXDBGVJZCEX-UHFFFAOYSA-N
triethylsilyl trifluoromethanesulfonate
2733308
Fuming Liquid
1.1600g/mL
Authentic
1.3890 to 1.3920
85.0°C to 86.0°C (12.0mmHg)
50g
99%
CF3SO3Si(C2H5)3
Solubility in water: soluble
CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F
264.34
264.34
99%
Videos
SDS Documents