missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetaldehyde ethyl hemiacetal, 90%, Thermo Scientific Chemicals

Catalog Number p-204641
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
433-27-2
C4H7F3O2
144.09
MFCD00000441
KLXJPQNHFFMLIG-UHFFFAOYSA-N
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol
9897
1-ethoxy-2,2,2-trifluoroethanol
CCOC(C(F)(F)F)O
This item is not returnable. View return policy
433-27-2
C4H7F3O2
144.09
MFCD00000441
KLXJPQNHFFMLIG-UHFFFAOYSA-N
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol
9897
1-ethoxy-2,2,2-trifluoroethanol
CCOC(C(F)(F)F)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.2400g/mL
Boiling Point 104°C to 105°C (745.0 mmHg)
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CF3CH(OC2H5)OH
Refractive Index 1.3410 to 1.344
Quantity 5 mL
Beilstein 01, IV, 3133
Specific Gravity 1.24
Formula Weight 144.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Trifluoroacetaldehyde ethyl hemiacetal, 90%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetaldehyde ethyl hemiacetal
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'